Learn More
Medchemexpress LLC 1-BOC-4-(2-Hydroxyethyl)piperazine-d4 | 1219802-10-4 | 99% | 234.33 | 25 MG
SDP
Catalog No. 5000403318
Encompass_Preferred
1-BOC-4-(2-Hydroxyethyl)piperazine-d4 is the deuterium labeled 1-BOC-4-(2-Hydroxyethyl)piperazine. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-03318 Supplier Medchemexpress LLC Supplier No. 25MGHY20797S
1-BOC-4-(2-Hydroxyethyl)piperazine-d4 is the deuterium labeled 1-BOC-4-(2-Hydroxyethyl)piperazine. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.