Learn More
Medchemexpress LLC 1-BOC-4-(2-Hydroxyethyl)piperazine-d4 | 1219802-10-4 | 99% | 234.33 | 50 MG
SDP
Catalog No. 5000400702
Encompass_Preferred
1-BOC-4-(2-Hydroxyethyl)piperazine-d4 is the deuterium labeled 1-BOC-4-(2-Hydroxyethyl)piperazine. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotope
Catalog No. 50-004-00702 Supplier Medchemexpress LLC Supplier No. HY20797S50MG
1-BOC-4-(2-Hydroxyethyl)piperazine-d4 is the deuterium labeled 1-BOC-4-(2-Hydroxyethyl)piperazine. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotope