Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Boc-4-(2-methoxycarbonylphenyl)piperazine, 97%, Thermo Scientific™

Catalog No. p-7052687
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH55302MD 250 mg
AAH5530203 1 g
AAH5530206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH55302MD Supplier Thermo Scientific Chemicals Supplier No. H55302MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870703-74-5
Molecular Formula C17H24N2O4
Molecular Weight (g/mol) 320.39
MDL Number MFCD07369760
InChI Key JFZGCXORLWXLGH-UHFFFAOYSA-N
Synonym 1-boc-4-2-methoxycarbonylphenyl piperazine, tert-butyl 4-2-methoxycarbonyl phenyl piperazine-1-carboxylate, methyl 2-boc-piperazin-1-yl benzoate, 1-boc-4-2-methoxycarbonylphenyl pipera, 1-boc-4-2-methoxycarbonyl phenyl-piperazine
PubChem CID 16217924
IUPAC Name tert-butyl 4-(2-methoxycarbonylphenyl)piperazine-1-carboxylate
SMILES COC(=O)C1=CC=CC=C1N1CCN(CC1)C(=O)OC(C)(C)C

Specifications

Color Yellow to Orange
Flash Point >110°C (230°F)
Refractive Index 1.532
Quantity 250 mg
UN Number UN2810
Formula Weight 320.39
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Boc-4-(2-methoxycarbonylphenyl)piperazine
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.