Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Boc-4-(2-nitrophenyl)piperazine, 97%, Thermo Scientific™

Catalog No. AAH63876MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63876MD 250 mg
AAH6387603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH63876MD Supplier Thermo Scientific Chemicals Supplier No. H63876MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170017-73-9
Molecular Formula C15H21N3O4
Molecular Weight (g/mol) 307.35
MDL Number MFCD08275017
InChI Key GTGIZVUOLBRDAO-UHFFFAOYSA-N
Synonym 1-boc-4-2-nitrophenyl piperazine, tert-butyl 4-2-nitrophenyl piperazine-1-carboxylate, 1-tert-butoxycarbonyl-4-2-nitrophenyl piperazine, 1-piperazinecarboxylic acid, 4-2-nitrophenyl-, 1,1-dimethylethyl ester, pubchem18863, acmc-209e0b, tert-butyl 4-2-nitrophenyl piperazinecarboxylate, tert-butyl 4-2-nitro phenyl piperazine-1-carboxylate, 1-1,1-dimethylethoxycarbony-4-2-nitrophenyl piperazine, 1-1,1-dimethylethoxycarbonyl-4-2-nitrophenyl piperazine
PubChem CID 21935210
IUPAC Name tert-butyl 4-(2-nitrophenyl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=CC=C1[N+]([O-])=O

Specifications

Quantity 250 mg
Formula Weight 307.34
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Boc-4-(2-nitrophenyl)piperazine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.