Learn More
Medchemexpress LLC 1-Boc-4-(4-aminobutyl)piperazine | 745048-07-1 | MFCD04038543 | 95.0% | 257.37 | C13H27N3O2 | 500 MG
SDP
Catalog No. 5000210147
Encompass_Preferred
1-Boc-4-(4-aminobutyl)piperazine is a Boc-protected piperazine derivative used as a drug intermediate in medicinal chemistry and small-molecule research applications.
  • Used as an intermediate in medicinal chemistry and small-molecule synthesis.
  • Boc-protected amine allows selective functionalization during multi-step syntheses.
  • Typical purity 95.0% is suitable for research and development workflows.
  • Available in multiple pack sizes, including 500 mg, for flexible experimental needs.
  • Store at 4°C and protect from light; in solvent, store at -80°C for up to 6 months or at -20°C for 1 month.
Catalog No. 50-002-10147 Supplier Medchemexpress LLC Supplier No. HYW045078500MG
1-Boc-4-(4-aminobutyl)piperazine is a Boc-protected piperazine derivative used as a drug intermediate in medicinal chemistry and small-molecule research applications.
  • Used as an intermediate in medicinal chemistry and small-molecule synthesis.
  • Boc-protected amine allows selective functionalization during multi-step syntheses.
  • Typical purity 95.0% is suitable for research and development workflows.
  • Available in multiple pack sizes, including 500 mg, for flexible experimental needs.
  • Store at 4°C and protect from light; in solvent, store at -80°C for up to 6 months or at -20°C for 1 month.