Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-(Boc-amino)-3-butene

Catalog No. 7402411G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-024-11G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 74-024-11G Supplier Sigma Aldrich Supplier No. 7402411G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-(Boc-amino)-3-butene

Specifications

CAS 156731-40-7
Density 0.929 g/mL (at 25°C)
Molecular Formula C9H17NO2
Refractive Index n20/D 1.443
Linear Formula C9H17NO2
MDL Number MFCD11975862
Quantity 100 g
Synonym tert-Butyl but-3-enylcarbamate; Boc-4-amino-1-butene
Molecular Weight (g/mol) 171.24
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.