Learn More
Medchemexpress LLC 1-Boc-azetidine-3-ca | 5g
SDP
Catalog No. 5000202322
Encompass_Preferred
1-Boc-azetidine-3-carboxylic acid is a non-cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs) 1-Boc-azetidine-3-carboxylic acid is also a alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs[2] /sup
Catalog No. 50-002-02322 Supplier Medchemexpress LLC Supplier No. HY401415G
1-Boc-azetidine-3-carboxylic acid is a non-cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs) 1-Boc-azetidine-3-carboxylic acid is also a alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs[2] /sup