Learn More
Medchemexpress LLC 1-Boc-azetidine-3-carboxylic acid | 142253-55-2 | MFCD01860897 | 98.3% | 201.22 g/mol
SDP
Catalog No. 5000203581
Encompass_Preferred
1-Boc-azetidine-3-carboxylic acid is a non-cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs) 1-Boc-azetidine-3-carboxylic acid is also a alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs[2] /sup
Catalog No. 50-002-03581 Supplier Medchemexpress LLC Supplier No. HY4014110G
1-Boc-azetidine-3-carboxylic acid is a non-cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs) 1-Boc-azetidine-3-carboxylic acid is also a alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs[2] /sup