Learn More
Medchemexpress LLC 1-Boc-azetidine-3-yl | 10g
SDP
Catalog No. 5000202201
Encompass_Preferred
1-Boc-azetidine-3-yl-methanol is a non-cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs) 1-Boc-azetidine-3-yl-methanol is also a alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs[1] /sup
Catalog No. 50-002-02201 Supplier Medchemexpress LLC Supplier No. HY4015210G
1-Boc-azetidine-3-yl-methanol is a non-cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs) 1-Boc-azetidine-3-yl-methanol is also a alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs[1] /sup