Learn More
Medchemexpress LLC 1-Boc-azetidine-3-yl |25g
SDP
Catalog No. 5000202874
Encompass_Preferred
1-Boc-azetidine-3-yl-methanol is a non-cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs) 1-Boc-azetidine-3-yl-methanol is also a alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs[1] /sup
Catalog No. 50-002-02874 Supplier Medchemexpress LLC Supplier No. HY4015225G
1-Boc-azetidine-3-yl-methanol is a non-cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs) 1-Boc-azetidine-3-yl-methanol is also a alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs[1] /sup