Learn More
Medchemexpress LLC 1-Boc-D-azetidine-2-carboxylic acid | 228857-58-7 | MFCD01320888 | >=97.0% | 201.22 g·mol⁻¹ | C9H15NO4 | 250 MG
SDP
Catalog No. 5000302386
Encompass_Preferred
1-Boc-D-azetidine-2-carboxylic acid is a Boc-protected D-azetidine amino acid derivative used as a chemical intermediate in medicinal chemistry and organic synthesis. It serves as a protected building block for introducing an azetidine unit into peptides and small molecules; intended for research use only.
  • Boc-protected azetidine scaffold for synthetic incorporation.
  • Suitable for peptide and small-molecule synthesis.
  • Available in multiple pack sizes for scale flexibility.
  • Typical purity ≥97% as supplied.
  • Molecular formula C9H15NO4; molecular weight 201.22 g·mol⁻¹.
Catalog No. 50-003-02386 Supplier Medchemexpress LLC Supplier No. 250MGHYW018751
1-Boc-D-azetidine-2-carboxylic acid is a Boc-protected D-azetidine amino acid derivative used as a chemical intermediate in medicinal chemistry and organic synthesis. It serves as a protected building block for introducing an azetidine unit into peptides and small molecules; intended for research use only.
  • Boc-protected azetidine scaffold for synthetic incorporation.
  • Suitable for peptide and small-molecule synthesis.
  • Available in multiple pack sizes for scale flexibility.
  • Typical purity ≥97% as supplied.
  • Molecular formula C9H15NO4; molecular weight 201.22 g·mol⁻¹.