Learn More
Medchemexpress LLC 1-Boc-D-azetidine-2-carboxylic acid | 228857-58-7 | MFCD01320888 | 99.0% | 201.22 | C9H15NO4 | 25 G
SDP
Catalog No. 5000302742
Encompass_Preferred
1-Boc-D-azetidine-2-carboxylic acid is a Boc-protected D-azetidine derivative used as a versatile building block in medicinal chemistry and small-molecule synthesis. It serves as an intermediate for preparation of biologically active compounds and is supplied with analytical documentation suitable for research and development use.
  • High purity (98.96%) with certificate of analysis available.
  • Boc-protected azetidine scaffold for selective synthetic transformations.
  • Available in multiple pack sizes, including bulk 25 g packaging.
  • Documented storage conditions for stability in solid and solution forms.
  • Accompanied by SDS and handling instructions for laboratory use.
Catalog No. 50-003-02742 Supplier Medchemexpress LLC Supplier No. 25GHYW018751
1-Boc-D-azetidine-2-carboxylic acid is a Boc-protected D-azetidine derivative used as a versatile building block in medicinal chemistry and small-molecule synthesis. It serves as an intermediate for preparation of biologically active compounds and is supplied with analytical documentation suitable for research and development use.
  • High purity (98.96%) with certificate of analysis available.
  • Boc-protected azetidine scaffold for selective synthetic transformations.
  • Available in multiple pack sizes, including bulk 25 g packaging.
  • Documented storage conditions for stability in solid and solution forms.
  • Accompanied by SDS and handling instructions for laboratory use.