Learn More
Medchemexpress LLC 1-Boc-D-azetidine-2-carboxylic acid | 228857-58-7 | MFCD01320888 | 99.0% | 201.22 g·mol⁻1 | C9H15NO4 | 10 G
SDP
Catalog No. 5000301029
Encompass_Preferred
1-Boc-D-azetidine-2-carboxylic acid is a Boc-protected azetidine building block used as a drug intermediate for synthesizing pharmaceutical and bioactive molecules. Supplied as a solid (CAS 228857-58-7; MW 201.22 g·mol⁻1), it is intended for organic synthesis and intermediate-scale reactions. Powder is stable at room temperature; solutions require low-temperature storage.
  • Boc protection enables selective functionalization
  • High purity suitable for synthesis and analytical use
  • Solid form for easy weighing and handling
  • Multiple package sizes available to support different scales
  • Storage guidance to preserve shelf life
Catalog No. 50-003-01029 Supplier Medchemexpress LLC Supplier No. HYW01875110G
1-Boc-D-azetidine-2-carboxylic acid is a Boc-protected azetidine building block used as a drug intermediate for synthesizing pharmaceutical and bioactive molecules. Supplied as a solid (CAS 228857-58-7; MW 201.22 g·mol⁻1), it is intended for organic synthesis and intermediate-scale reactions. Powder is stable at room temperature; solutions require low-temperature storage.
  • Boc protection enables selective functionalization
  • High purity suitable for synthesis and analytical use
  • Solid form for easy weighing and handling
  • Multiple package sizes available to support different scales
  • Storage guidance to preserve shelf life