Learn More
Medchemexpress LLC 1-Boc-D-azetidine-2-carboxylic acid | 228857-58-7 | MFCD01320888 | 99.0% | 201.22 g/mol | C9H15NO4 | 5 G
SDP
Catalog No. 5000301082
Encompass_Preferred
1-Boc-D-azetidine-2-carboxylic acid is a Boc-protected D-azetidine amino acid derivative used as a synthetic intermediate in medicinal chemistry and organic synthesis. The N-Boc protection facilitates selective amine protection and subsequent coupling or derivatization of the azetidine ring, enabling preparation of azetidine-containing active compounds and building blocks.
  • Boc-protected azetidine scaffold for versatile synthetic transformations.
  • High purity (≈99.0%).
  • Molecular weight 201.22 g/mol; formula C9H15NO4.
  • Available in multiple small-scale pack sizes for laboratory synthesis.
  • Stable as a solid at room temperature for up to 3 years; in solvent, long-term storage at -80°C.
  • Suitable as a building block for medicinal chemistry and lead optimization.
Catalog No. 50-003-01082 Supplier Medchemexpress LLC Supplier No. HYW0187515G
1-Boc-D-azetidine-2-carboxylic acid is a Boc-protected D-azetidine amino acid derivative used as a synthetic intermediate in medicinal chemistry and organic synthesis. The N-Boc protection facilitates selective amine protection and subsequent coupling or derivatization of the azetidine ring, enabling preparation of azetidine-containing active compounds and building blocks.
  • Boc-protected azetidine scaffold for versatile synthetic transformations.
  • High purity (≈99.0%).
  • Molecular weight 201.22 g/mol; formula C9H15NO4.
  • Available in multiple small-scale pack sizes for laboratory synthesis.
  • Stable as a solid at room temperature for up to 3 years; in solvent, long-term storage at -80°C.
  • Suitable as a building block for medicinal chemistry and lead optimization.