Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Boc-homopiperazine | 112275-50-0 | 99.87% | 200.28 | 50 G
SDP

Catalog No. 5000401460
Encompass_Preferred

1-Boc-homopiperazine is a drug intermediate used for the synthesis of bacterial FtsZ inhibitors. It is a colorless to light yellow liquid for research use only.

  • Used for the synthesis of bacterial FtsZ inhibitors
  • Colorless to light yellow liquid appearance
  • High purity of 99.87%
  • Molecular weight of 200.28
  • Recommended storage at 4°C, protected from light

Catalog No. 50-004-01460 Supplier Medchemexpress LLC Supplier No. HY4018150G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Boc-homopiperazine is a drug intermediate used for the synthesis of bacterial FtsZ inhibitors. It is a colorless to light yellow liquid for research use only.

  • Used for the synthesis of bacterial FtsZ inhibitors
  • Colorless to light yellow liquid appearance
  • High purity of 99.87%
  • Molecular weight of 200.28
  • Recommended storage at 4°C, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.