Learn More
Medchemexpress LLC 1-Boc-homopiperazine | 112275-50-0 | MFCD00276987 | 99.9% | 200.28 | 100 G
SDP
Catalog No. 5000404075
Encompass_Preferred
1-Boc-homopiperazine is a drug intermediate that can be used for the synthesis of bacterial FtsZ inhibitors.
  • Used for the synthesis of bacterial FtsZ inhibitors.
Catalog No. 50-004-04075 Supplier Medchemexpress LLC Supplier No. HY40181100G
1-Boc-homopiperazine is a drug intermediate that can be used for the synthesis of bacterial FtsZ inhibitors.
  • Used for the synthesis of bacterial FtsZ inhibitors.