missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 1-Boc-piperidine-4-carboxaldehyde

Catalog No. 722022500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-202-2500MG 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 72-202-2500MG Supplier Sigma Aldrich Supplier No. 722022500MG
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Boc-piperidine-4-carboxaldehyde

Specifications

CAS 137076-22-3
Melting Point 34°C to 40°C
Molecular Formula C11H19NO3
Linear Formula C11H19NO3
MDL Number MFCD02179019
Quantity 1 g
Synonym 4-Formylcyclohexanecarboxylic acid tert-butyl ester; tert-Butyl 4-formylpiperidine-1-carboxylate
Molecular Weight (g/mol) 213.27
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.