Learn More
Accela Chembio Inc 1-boc-pyrazole | 5g | 219580-32-2 | MFCD00800729 | 97+% | D: 1.036 | Shelf Life: 1620 Days | Light Sensitive/+4
Supplier:  Accela Chembio Inc SY0040855G
Encompass_Preferred
1-boc-pyrazole | 5g | 219580-32-2 | MFCD00800729 | 97+% | D: 1.036 | Shelf Life: 1620 Days | Light Sensitive/+4
Catalog No. 50-250-373