Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2,3,5,6-tetramethylbenzene, 99%

Catalog No. AAL0146514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0146514 25 g
AAL0146506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0146514 Supplier Thermo Scientific Chemicals Supplier No. L0146514
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1646-53-3 | C10H13Br | 213.118 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1646-53-3
Molecular Formula C10H13Br
Molecular Weight (g/mol) 213.118
MDL Number MFCD00000070
InChI Key WJKBPTLQJXKEHC-UHFFFAOYSA-N
Synonym bromodurene, 3-bromodurene, 1-bromo-2,3,5,6-tetramethylbenzene, benzene, 3-bromo-1,2,4,5-tetramethyl, bromdurol, duryl bromide, bromodurene, pract., acmc-1bz04, 2,3,5,6-tetramethylbromobenzene, 1-bromo-2,5,6-tetramethylbenzene
PubChem CID 74240
IUPAC Name 3-bromo-1,2,4,5-tetramethylbenzene
SMILES CC1=CC(=C(C(=C1C)Br)C)C

Specifications

Melting Point 58°C to 62°C
Boiling Point 115°C to 120°C (5 mmHg)
Quantity 25 g
Beilstein 2207158
Formula Weight 213.12
Percent Purity 99%
Chemical Name or Material 1-Bromo-2,3,5,6-tetramethylbenzene
EINECSNumber 216-707-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.