Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2,4,5-trifluorobenzene, 98%, Thermo Scientific™

Catalog No. AC271160050
Change view
Click to view available options
Glass bottle; 5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC271160050 Glass bottle; 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC271160050 Supplier Thermo Scientific Chemicals Supplier No. 271160050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Specifications

CAS 327-52-6
CAS Min % 98.5
CAS Max % 100.0
Density 1.8020g/mL
Boiling Point 143.0°C to 144.0°C
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H2BrF3
Refractive Index 1.4850 to 1.4870
Linear Formula BrC6H2F3
Quantity 5g
MDL Number MFCD00000306
Beilstein 05, 679
Specific Gravity 1.802
Synonym 2,4,5-trifluorobromobenzene, 2-bromo-1,4,5-trifluorobenzene, benzene, 1-bromo-2,4,5-trifluoro, 1-bromo-2,4,5-trifluoro-benzene, paragos 390205, 5-bromo-1,2,4-trifluorobenzene, fr bf df ee, pubchem2306, acmc-1cs6j, ksc224g3h
Solubility Information Solubility in water: insoluble.
InChI Key DVTULTINXNWGJY-UHFFFAOYSA-N
SMILES C1=C(C(=CC(=C1F)Br)F)F
IUPAC Name 1-bromo-2,4,5-trifluorobenzene
Molecular Weight (g/mol) 210.98
PubChem CID 67597
Formula Weight 210.98
Percent Purity 98%
Chemical Name or Material 1-Bromo-2, 4, 5-trifluorobenzene, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea
EINECSNumber 206-318-8
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.