Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Bromo-2,4,6-triisopropylbenzene

Catalog No. 6379395G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-793-95G 5 g
63-793-925G 25 g
63-793-9100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 63-793-95G Supplier Sigma Aldrich Supplier No. 6379395G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-bromo-2,4,6-triisopropylbenzene can be used in the synthesis of 1-fluoro-2,4,6-triisopropylbenzene, a fluorine-substituted aromatic building block for agrochemical synthesis and pharmaceutical production
  • It can also be used in the synthesis of di-2,4,6-triisopropylphenyl diselenide via reaction with metallic selenium in the presence of tertiary-butyl lithium

Specifications

CAS 21524-34-5
Density 1.118 g/mL (at 25°C (literature))
Boiling Point 96°C to 97°C (0.1 mmHg)
Molecular Formula C15H23Br
Refractive Index n20/D 1.523 (literature)
Linear Formula C15H23Br
MDL Number MFCD00051547
Quantity 5 g
Synonym 2,4,6-Triisopropylbromobenzene; 2-Bromo-1,3,5-triisopropylbenzene
Molecular Weight (g/mol) 283.25
Percent Purity 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.