Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2,4-difluoro-5-nitrobenzene 98.0+%, TCI America™
SDP

Catalog No. B56885G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56885G 5 g
B56881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56885G Supplier TCI America Supplier No. B56885G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 345-24-4
Molecular Formula C6H2BrF2NO2
Molecular Weight (g/mol) 237.99
MDL Number MFCD00129166
InChI Key OOUUWURPSUSDTD-UHFFFAOYSA-N
Synonym 5-Bromo-2,4-difluoronitrobenzene
PubChem CID 223078
IUPAC Name 1-bromo-2,4-difluoro-5-nitrobenzene
SMILES [O-][N+](=O)C1=CC(Br)=C(F)C=C1F

Specifications

Melting Point 19°C
Color Yellow
Boiling Point 97°C
Quantity 5 g
Formula Weight 237.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-2,4-difluoro-5-nitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.