Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2,4-dinitrobenzene, MP Biomedicals

Catalog No. ICN202344 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN202344 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN202344 Supplier MP Biomedicals, Inc Supplier No. 0520234410
Add to Cart
Add to Cart

Chemical Identifiers

CAS 584-48-5
Molecular Formula C6H3BrN2O4
Molecular Weight (g/mol) 247.004
InChI Key PBOPJYORIDJAFE-UHFFFAOYSA-N
Synonym 2,4-dinitrobromobenzene, 4-bromo-1,3-dinitrobenzene, o,p-dinitrophenyl bromide, 2,4-dinitrophenyl bromide, dnbb, benzene, 1-bromo-2,4-dinitro, 2, 4-dinitrobromobenzene, 1-bromo-2,4-dinitrobenze, unii-4ms3ftw380, ccris 1801
PubChem CID 11441
ChEBI CHEBI:58999
IUPAC Name 1-bromo-2,4-dinitrobenzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])Br

Specifications

Quantity 10 g
Formula Weight 247.01
Chemical Name or Material 1-Bromo-2, 4-dinitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.