Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2-butanone, tech. 90%, stab. with calcium carbonate, Thermo Scientific Chemicals

Catalog No. AAL1208103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1208103 1 g
AAL1208106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1208103 Supplier Thermo Scientific Chemicals Supplier No. L1208103
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 816-40-0
Molecular Formula C4H7BrO
Molecular Weight (g/mol) 151.003
MDL Number MFCD00000207
InChI Key CCXQVBSQUQCEEO-UHFFFAOYSA-N
Synonym 1-bromo-2-butanone, 2-butanone, 1-bromo, 1-bromobutanone, bromomethyl ethyl ketone, bromobutanone, bromobutan-2-one, l-bromo-2-butanone, acmc-20aoxd, 1-bromo-butan-2-one
PubChem CID 13156
IUPAC Name 1-bromobutan-2-one
SMILES CCC(=O)CBr

Specifications

Density 1.479
Boiling Point 48°C to 49°C (10 mmHg)
Flash Point 68°C (154°F)
Odor Pungent
Refractive Index 1.465
Quantity 1 g
UN Number UN2927
Beilstein 741894
Formula Weight 151.01
Percent Purity 85%
Grade Technical
Chemical Name or Material 1-Bromo-2-butanone, stabilized with calcium carbonate
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H330-H314-H318-H227
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 212-431-3
RTECSNumber EL7000000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.