Learn More
Medchemexpress LLC 1-Bromo-2-chlorobenzene-d4 | 1219795-51-3 | 195.48 | 50 MG
SDP
Catalog No. 5000449252
Encompass_Preferred
1-Bromo-2-chlorobenzene-d4 is the deuterium labeled 1-Bromo-2-chlorobenzene. It is for research use only.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-49252 Supplier Medchemexpress LLC Supplier No. HYW004051S50MG
1-Bromo-2-chlorobenzene-d4 is the deuterium labeled 1-Bromo-2-chlorobenzene. It is for research use only.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.