Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific 1-Bromo-2-fluorobenzene, 1072-85-1, MFCD00000282, 100g

Catalog No. 500113334
Encompass_Preferred

Molecular Formula C6H4BrF, Purity 99%, Molecular Weight 175.01, Boiling Point 156

Catalog No. 50-011-3334 Supplier Matrix Scientific Supplier No. 001728100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula C6H4BrF, Purity 99%, Molecular Weight 175.01, Boiling Point 156

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.