Learn More
Medchemexpress LLC 1-bromo-2-fluorobenzene-d4 | 50592-35-3 | 99.0% | 179.02 | C6D4BrF | 10 MG
SDP
Catalog No. 5000298340
Encompass_Preferred
1-Bromo-2-fluorobenzene-d4 is the deuterium-labeled analogue of 1-bromo-2-fluorobenzene intended for use as a tracer and as an internal standard in quantitative analytical techniques such as NMR, GC-MS, and LC-MS. It is supplied in small, laboratory-scale quantities with high isotopic and chemical purity for analytical applications.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS analytical methods.
  • High chemical purity (99.0%) to ensure reliable results.
  • Available in small, controlled quantities for laboratory use.
  • Liquid form with documented density for handling and formulation.
  • Recommended storage conditions provided to preserve stability.
Catalog No. 50-002-98340 Supplier Medchemexpress LLC Supplier No. HY42397S10MG
1-Bromo-2-fluorobenzene-d4 is the deuterium-labeled analogue of 1-bromo-2-fluorobenzene intended for use as a tracer and as an internal standard in quantitative analytical techniques such as NMR, GC-MS, and LC-MS. It is supplied in small, laboratory-scale quantities with high isotopic and chemical purity for analytical applications.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS analytical methods.
  • High chemical purity (99.0%) to ensure reliable results.
  • Available in small, controlled quantities for laboratory use.
  • Liquid form with documented density for handling and formulation.
  • Recommended storage conditions provided to preserve stability.