Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2-iodobenzene (stabilized with Copper chip) 98.0+%, TCI America™
SDP

Catalog No. B112625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B112625G 25 g
B11265G 5 g
B1126250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B112625G Supplier TCI America Supplier No. B112625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 583-55-1
Molecular Formula C6H4BrI
Molecular Weight (g/mol) 282.906
MDL Number MFCD00001030
InChI Key OIRHKGBNGGSCGS-UHFFFAOYSA-N
Synonym 2-bromoiodobenzene, o-bromoiodobenzene, 2-bromo-iodobenzene, benzene, 1-bromo-2-iodo, bromoiodobenzene, 1-bromo-2-iodo-benzene, attercop-chm at127403, chemwish ic03500, iodobromobenzene, 2-iodobromobenzene
PubChem CID 11415
IUPAC Name 1-bromo-2-iodobenzene
SMILES C1=CC=C(C(=C1)Br)I

Specifications

Boiling Point 120°C
Quantity 25 g
Formula Weight 282.91
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-iodobenzene (stabilized with Copper chip)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.