Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Bromo-2-methyl-1-propene

Catalog No. 27714250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-714-250G 50 g
27-714-210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 27-714-250G Supplier Sigma Aldrich Supplier No. 27714250G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-bromo-2-methyl-1-propene has been used in:
  • Preparation of esters of 2,4-dienoic acids via palladium catalyzed reaction with methyl acrylate in the presence of triethylamine
  • Total synthesis of ocular age pigment, a2-e
  • Synthesis of 3-nor-geraniol [2h2]may darken in storage

Specifications

CAS 3017-69-4
Density 1.318 g/mL (at 25°C (literature))
Boiling Point 92°C (lit.)
Molecular Formula C4H7Br
Refractive Index n20/D 1.462 (literature)
Linear Formula (CH3)2 C=CHBr
MDL Number MFCD00009885
Quantity 50 g
Synonym Isocrotyl bromide
Molecular Weight (g/mol) 135
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.