Learn More
Medchemexpress LLC 1-bromo-2-methyl-3-nitrobenzene | 55289-35-5 | MFCD00009792 | 99.5% | 216.03 g/mol | C7H6BrNO2 | 25 G
SDP
Catalog No. 5000303008
Encompass_Preferred
1-Bromo-2-methyl-3-nitrobenzene is an aromatic organic intermediate used in medicinal and synthetic chemistry. Its bromine substituent serves as a leaving group for cross-coupling and nucleophilic substitution reactions, while the nitro group provides electron-withdrawing directing effects. It is commonly used as a precursor for constructing more complex aromatic scaffolds.
  • Enables cross-coupling and substitution reactions
  • Nitro group provides electron-withdrawing directing effects
  • High purity suitable for research applications
  • Supplied in multiple package sizes for scalability
  • Stable when stored under recommended conditions
Catalog No. 50-003-03008 Supplier Medchemexpress LLC Supplier No. 25GHYW014159
1-Bromo-2-methyl-3-nitrobenzene is an aromatic organic intermediate used in medicinal and synthetic chemistry. Its bromine substituent serves as a leaving group for cross-coupling and nucleophilic substitution reactions, while the nitro group provides electron-withdrawing directing effects. It is commonly used as a precursor for constructing more complex aromatic scaffolds.
  • Enables cross-coupling and substitution reactions
  • Nitro group provides electron-withdrawing directing effects
  • High purity suitable for research applications
  • Supplied in multiple package sizes for scalability
  • Stable when stored under recommended conditions