Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Bromo-2-methylnaphthalene

Catalog No. 124109100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-410-9100G 100 g
12-410-925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-410-9100G Supplier Sigma Aldrich Supplier No. 124109100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Dynamic phosphorescence quenching of 1-bromo-2-methylnaphthalene without deoxygenation has been used for selective sensing of cu(ii) at ngml-1 levels
  • It undergoes asymmetric cross-coupling reaction with its corresponding grignard reagent using a nickel catalyst to form non-racemic 2,2α-dimethyl-1,1α-binaphthyl.1-bromo-2-methylnaphthalene was used as test compound in determination of halogenated hydrocarbons at trace levels by supercritical fluid chromatography-microwave-induced plasma mass spectrometry

Specifications

CAS 2586-62-1
Density 1.418 g/mL (at 25°C (literature))
Boiling Point 296°C (lit.)
Molecular Formula C11H9Br
Refractive Index n20/D 1.648 (literature)
Linear Formula CH3C10H6Br
MDL Number MFCD00003871
Quantity 100 g
Molecular Weight (g/mol) 221.09
Percent Purity 90%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.