Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2-nitrobenzene, 99%

Catalog No. p-7081637
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1168618 50 g
AAA1168630 250 g
AAA116860B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1168618 Supplier Thermo Scientific Chemicals Supplier No. A1168618
Only null left
Add to Cart

CAS: 577-19-5 | C6H4BrNO2 | 202.007 g/mol

1-Bromo-2-nitrobenzene is used in organic synthesis. It also can react with 2-methylamino-benzoic acid to get N-methyl-N-(2-nitro-phenyl)-anthranilic acid. And it is also used in the preparation of 4-methoxy-2?-nitrodiphenyl ether, 1-methoxy-3,5-bis-(2-nitro-phenoxy)benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-nitrobenzene is used in organic synthesis. It also can react with 2-methylamino-benzoic acid to get N-methyl-N-(2-nitro-phenyl)-anthranilic acid. And it is also used in the preparation of 4-methoxy-2′-nitrodiphenyl ether, 1-methoxy-3,5-bis-(2-nitro-phenoxy)benzene.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 577-19-5
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.007
MDL Number MFCD00007045
InChI Key ORPVVAKYSXQCJI-UHFFFAOYSA-N
Synonym 2-bromonitrobenzene, o-bromonitrobenzene, o-nitrobromobenzene, 2-nitrobromobenzene, benzene, 1-bromo-2-nitro, o-nitrophenyl bromide, 2-bromo-1-nitrobenzene, 1-bromo-2-nitro-benzene, 2-bromo nitrobenzene, benzene, bromonitro
PubChem CID 11341
IUPAC Name 1-bromo-2-nitrobenzene
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])Br

Specifications

Melting Point 40°C to 44°C
Boiling Point 261°C
Flash Point 112°C (233°F)
Quantity 50 g
UN Number UN3459
Beilstein 2044109
Solubility Information Insoluble in water.
Formula Weight 202.01
Percent Purity 99%
Chemical Name or Material 1-Bromo-2-nitrobenzene
Hazard Category H302+H312+H332-H315-H319-H335-H373
Hazard Statement GHS H Statement
H331-H373-H302-H312-H315-H319-H335
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROBROMOBENZENES, SOLID
EINECSNumber 209-409-0
RTECSNumber CY9040450
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.