Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-bromo-3,4-difluorobenzene-d3 | 1219799-14-0 | 98.6% | 196.01 g/mol | C6D3BrF2 | 25 MG
SDP

Catalog No. 5000303390
Encompass_Preferred

1-Bromo-3,4-difluorobenzene-d3 is a deuterium-labeled analogue of 1-bromo-3,4-difluorobenzene supplied as an isotope-labeled reference standard for analytical and research applications. It is provided with batch-specific quality documentation and handling information to support use as an internal standard or synthetic intermediate in studies requiring deuterated material.

  • Deuterium-labeled internal standard for mass spectrometry and NMR.
  • High chemical purity: 98.6% (assay) and high isotopic enrichment.
  • Molecular formula C6D3BrF2; molecular weight 196.01 g/mol.
  • Colorless to light yellow liquid suitable for solution-phase workflows.
  • Certificate of analysis and safety data sheet available for batch-specific details.
  • Recommended storage at -20°C in pure form to preserve stability.

Catalog No. 50-003-03390 Supplier Medchemexpress LLC Supplier No. 25MGHYW015079S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

1-Bromo-3,4-difluorobenzene-d3 is a deuterium-labeled analogue of 1-bromo-3,4-difluorobenzene supplied as an isotope-labeled reference standard for analytical and research applications. It is provided with batch-specific quality documentation and handling information to support use as an internal standard or synthetic intermediate in studies requiring deuterated material.

  • Deuterium-labeled internal standard for mass spectrometry and NMR.
  • High chemical purity: 98.6% (assay) and high isotopic enrichment.
  • Molecular formula C6D3BrF2; molecular weight 196.01 g/mol.
  • Colorless to light yellow liquid suitable for solution-phase workflows.
  • Certificate of analysis and safety data sheet available for batch-specific details.
  • Recommended storage at -20°C in pure form to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.