Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-3,5-difluorobenzene, 98%

Catalog No. AAA1405314
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1405314 25 g
AAA1405306 5 g
AAA1405322 100 g
AAA1405330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1405314 Supplier Thermo Scientific Chemicals Supplier No. A1405314
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 461-96-1 | C6H3BrF2 | 192.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 461-96-1
Molecular Formula C6H3BrF2
Molecular Weight (g/mol) 192.99
MDL Number MFCD00010305
InChI Key JHLKSIOJYMGSMB-UHFFFAOYSA-N
Synonym 3,5-difluorobromobenzene, 1-bromo-3,5-difluoro-benzene, 1-bromo-3,5-difluoro benzene, benzene, 1-bromo-3,5-difluoro, 3,5-difluoro-bromobenzene, pubchem1042, acmc-1ae0k, 3,5-difluoro bromobenzene, bromo-3,5-difluorobenzene, 3,5-difluorophenyl bromide
PubChem CID 136313
IUPAC Name 1-bromo-3,5-difluorobenzene
SMILES FC1=CC(Br)=CC(F)=C1

Specifications

Melting Point -27°C
Density 1.676
Boiling Point 140°C to 141°C
Flash Point 44°C (111°F)
Refractive Index 1.499
Quantity 25 g
UN Number UN1993
Beilstein 1931520
Formula Weight 192.99
Percent Purity 98%
Chemical Name or Material 1-Bromo-3,5-difluorobenzene
Show More Show Less
Hazard Category H226-H302-H315-H317-H319-H373
Hazard Statement GHS H Statement
H226-H373-H302-H315-H317
Flammable liquid and vapour.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P272-P280-P301+P312-P303+P361+P353-P305+P351+P338-P314-P330-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 416-710-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.