Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Bromo-3,5-dimethylbenzene

Catalog No. 27631625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-631-625G 25 g
27-631-6100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 27-631-625G Supplier Sigma Aldrich Supplier No. 27631625G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Palladium catalyzed carbon-oxygen coupling of 1-bromo-3,5-dimethylbenzene and o-cresol to potassium hydroxide to produce o-tolyl-3,5-xylyl ether has been reported.1-Bromo-3,5-dimethylbenzene has been used in the synthesis of substituted 2,2α-bis(diphenylphosphanylmethyl)-1,1α-binaphthyl derivatives.

Specifications

CAS 556-96-7
Density 1.362 g/mL (at 25°C (literature))
Boiling Point 202°C to 204°C (lit.)
Molecular Formula C8H9Br
Refractive Index n20/D 1.549 (literature)
Linear Formula (CH3)2C6H3Br
MDL Number MFCD00000087
Quantity 25 g
Synonym 5-Bromo-m-xylene
Molecular Weight (g/mol) 185.06
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.