Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-3-chloro-5-fluorobenzene, 98%, Thermo Scientific™

Catalog No. AC448750050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC448750050 5 g
AC448750250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC448750050 Supplier Thermo Scientific Chemicals Supplier No. 448750050

Chemical Identifiers

CAS 33863-76-2
Molecular Formula C6H3BrClF
Molecular Weight (g/mol) 209.45
InChI Key GGMDFPMASIXEIR-UHFFFAOYSA-N
Synonym 3-chloro-5-fluorobromobenzene, 1-bromo3-chloro-5-fluorobenzene, 1-bromo-3-chloro-5-fluoro-benzene, benzene, 1-bromo-3-chloro-5-fluoro, 1,3,5-fluorochlorobromobenzene, pubchem2261, acmc-209i3i, ksc494q7t, 3-chloro-5-fluoro-bromobenzene, 1-bromo-3-chloro-5-flurobenzene
PubChem CID 2736223
IUPAC Name 1-bromo-3-chloro-5-fluorobenzene
SMILES C1=C(C=C(C=C1Cl)Br)F

Specifications

Density 1.7200g/mL
Boiling Point 72°C (5.0 mmHg)
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H3(Cl)F
Quantity 5 g
Specific Gravity 1.72
Formula Weight 209.45
Percent Purity 98%
Chemical Name or Material 1-Bromo-3-chloro-5-fluorobenzene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.