Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Bromo-3-chloropropane

Catalog No. 501859486
Encompass
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-185-9486 1 kg
50-185-9487 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-185-9486 Supplier Sigma Aldrich Supplier No. B624041KG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-bromo-3-chloropropane is generally used as c3 linker/spacer in a wide range of molecules including medicinally important compounds. some of the examples are:
  • Synthesis of polymer nanoparticles (nps) to deliver doxorubicin, camptothecin, and cisplatin
  • Synthesis of antiprion active acridine derivatives for prion infection
  • Total synthesis of aspidospermine, aspidospermidine, and quebrachamine and (+/-)-crispine a
TRUSTED_SUSTAINABILITY

Specifications

CAS 109-70-6
Density 1.592 g/mL (at 25°C (literature))
Boiling Point 144°C to 145°C (lit.)
Molecular Formula C3H6BrCl
Refractive Index n20/D 1.486 (literature)
Linear Formula Cl(CH2)3 Br
Quantity 1 kg
MDL Number MFCD00000998
Synonym I-BCP; Trimethylene bromochloride; Trimethylene chlorobromide
Molecular Weight (g/mol) 157.44
Percent Purity 99%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.