Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-3-(difluoromethoxy)benzene 98.0+%, TCI America™
SDP

Catalog No. B41895G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B41895G Supplier TCI America Supplier No. B41895G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 262587-05-3
Molecular Formula C7H5BrF2O
Molecular Weight (g/mol) 223.017
MDL Number MFCD00236233
InChI Key NWBDGKNKAFGQQL-UHFFFAOYSA-N
Synonym 1-bromo-3-difluoromethoxy benzene, 3-difluoromethoxy bromobenzene, benzene, 1-bromo-3-difluoromethoxy, 1-difluoromethoxy-3-bromobenzene, pubchem18537, acmc-1crzp, 3-difluoromethoxybromobenzene, ksc493c1b, 3-bromo-1-difluoromethoxy benzene, 3-difluoromethyl bromobenzene
PubChem CID 2737006
IUPAC Name 1-bromo-3-(difluoromethoxy)benzene
SMILES C1=CC(=CC(=C1)Br)OC(F)F

Specifications

Color Yellow
Boiling Point 197°C
Quantity 5 g
Formula Weight 223.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-3-(difluoromethoxy)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.