Learn More
Medchemexpress LLC 1-Bromo-3-iodobenzene | 591-18-4 | MFCD00001043 | 99.99% | 282.91 | 250 G
SDP
Catalog No. 5000403101
Encompass_Preferred
1-Bromo-3-iodobenzene is a biochemical reagent and an important organic synthetic intermediate. It can be used for the synthesis of other active compounds. It acts as a dihaloarene substrate in the palladium-catalyzed three-component tandem reaction, enabling the one-pot synthesis of unsymmetrical biaryl thioglycosides with thiosugars and arylboronic acids via the selective formation of S-C and C-C bonds for efficient transformation.
  • Can be used for the synthesis of other active compounds.
  • Functions as a dihaloarene substrate in palladium-catalyzed three-component tandem reactions.
  • Enables one-pot synthesis of unsymmetrical biaryl thioglycosides.
Catalog No. 50-004-03101 Supplier Medchemexpress LLC Supplier No. 250GHY32149
Add to Cart
Add to Cart
1-Bromo-3-iodobenzene is a biochemical reagent and an important organic synthetic intermediate. It can be used for the synthesis of other active compounds. It acts as a dihaloarene substrate in the palladium-catalyzed three-component tandem reaction, enabling the one-pot synthesis of unsymmetrical biaryl thioglycosides with thiosugars and arylboronic acids via the selective formation of S-C and C-C bonds for efficient transformation.
  • Can be used for the synthesis of other active compounds.
  • Functions as a dihaloarene substrate in palladium-catalyzed three-component tandem reactions.
  • Enables one-pot synthesis of unsymmetrical biaryl thioglycosides.