Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-3-iodobenzene, 98+%, Thermo Scientific™

Catalog No. AC249420250
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC249420250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC249420250 Supplier Thermo Scientific Chemicals Supplier No. 249420250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-18-4
Molecular Formula C6H4BrI
Molecular Weight (g/mol) 282.91
MDL Number MFCD00001043
InChI Key CTPUUDQIXKUAMO-UHFFFAOYSA-N
Synonym 3-bromoiodobenzene, benzene, 1-bromo-3-iodo, m-bromoiodobenzene, 1-bromo-3-iodo-benzene, 3-bromo-1-iodobenzene, attercop-chm at127411, m-bromjodbenzol, 3-iodobromobenzene, 3-bromo iodobenzene
PubChem CID 11561
IUPAC Name 1-bromo-3-iodobenzene
SMILES BrC1=CC=CC(I)=C1

Specifications

Melting Point -9.3°C
Density 2.2100g/mL
Boiling Point 120.0°C (18.0 mmHg)
Flash Point >110°C
Refractive Index 1.6590 to 1.6615
Linear Formula BrC6H4I
Quantity 25 mL
Specific Gravity 2.21
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 282.89
Chemical Name or Material 1-Bromo-3-iodobenzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.