Learn More
Medchemexpress LLC 1-bromo-3-methoxybenzene-d3 | 873928-72-4 | >96.0% | 190.05 | C7H4D3BrO | 250 MG
SDP
Catalog No. 5000302350
Encompass_Preferred
1-Bromo-3-methoxybenzene-d3 is the deuterium-labeled analogue of 1-bromo-3-methoxybenzene (CAS 873928-72-4). It is provided in milligram-scale quantities as an isotopically labeled reagent for analytical chemistry and metabolic tracing applications, enabling precise tracking of molecular fragments in mass spectrometry and kinetic studies.
  • Deuterium-labeled reagent for tracer and metabolic studies.
  • Suitable as an internal standard for mass spectrometry analyses.
  • Available in milligram-scale quantities (100 mg-1 g).
  • Molecular formula C7H4D3BrO; molecular weight 190.05 g/mol.
  • Intended for research use in analytical and preparative workflows.
Catalog No. 50-003-02350 Supplier Medchemexpress LLC Supplier No. 250MGHYW016152S
Add to Cart
Add to Cart
1-Bromo-3-methoxybenzene-d3 is the deuterium-labeled analogue of 1-bromo-3-methoxybenzene (CAS 873928-72-4). It is provided in milligram-scale quantities as an isotopically labeled reagent for analytical chemistry and metabolic tracing applications, enabling precise tracking of molecular fragments in mass spectrometry and kinetic studies.
  • Deuterium-labeled reagent for tracer and metabolic studies.
  • Suitable as an internal standard for mass spectrometry analyses.
  • Available in milligram-scale quantities (100 mg-1 g).
  • Molecular formula C7H4D3BrO; molecular weight 190.05 g/mol.
  • Intended for research use in analytical and preparative workflows.