Learn More
Medchemexpress LLC 1-bromo-3-methoxybenzene-d3 | 873928-72-4 | MFCD12760295 | >98.0% | 190.05 g/mol | C7H4D3BrO | 500 MG
SDP
Catalog No. 5000303220
Encompass_Preferred
1-Bromo-3-methoxybenzene-d3 is the deuterium-labeled analogue of 1-bromo-3-methoxybenzene supplied for research use. It serves as an isotopically labeled reagent and internal standard for analytical and synthetic workflows, particularly in studies that require precise atom tracking and quantitation.
  • Deuterium-labeled (d3) isotopic analogue.
  • Useful as an internal standard for NMR, GC-MS, and LC-MS.
  • Suitable for metabolic tracing and isotopic labelling studies.
  • Available in small pack sizes appropriate for laboratory research.
  • Molecular formula C7H4D3BrO; molecular weight 190.05 g/mol.
Catalog No. 50-003-03220 Supplier Medchemexpress LLC Supplier No. HYW016152S500MG
1-Bromo-3-methoxybenzene-d3 is the deuterium-labeled analogue of 1-bromo-3-methoxybenzene supplied for research use. It serves as an isotopically labeled reagent and internal standard for analytical and synthetic workflows, particularly in studies that require precise atom tracking and quantitation.
  • Deuterium-labeled (d3) isotopic analogue.
  • Useful as an internal standard for NMR, GC-MS, and LC-MS.
  • Suitable for metabolic tracing and isotopic labelling studies.
  • Available in small pack sizes appropriate for laboratory research.
  • Molecular formula C7H4D3BrO; molecular weight 190.05 g/mol.