Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-3-methyl-2-butene (stabilized with Silver chip) 90.0+%, TCI America™
SDP

Catalog No. B084425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B084425G 25 g
B08445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B084425G Supplier TCI America Supplier No. B084425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 870-63-3
Molecular Formula C5H9Br
Molecular Weight (g/mol) 149.031
MDL Number MFCD00000242
InChI Key LOYZVRIHVZEDMW-UHFFFAOYSA-N
Synonym 1-bromo-3-methyl-2-butene, 3,3-dimethylallyl bromide, prenyl bromide, 3-methyl-2-butenyl bromide, 2-butene, 1-bromo-3-methyl, dimethylallyl bromide, 1-bromo-3-methyl-but-2-ene, .gamma.,.gamma.-dimethylallyl bromide, 3-methylcrotyl bromide, 3,3-dimethylallylbromide
PubChem CID 70092
IUPAC Name 1-bromo-3-methylbut-2-ene
SMILES CC(=CCBr)C

Specifications

Color Yellow
Boiling Point 135°C
Quantity 25 g
UN Number 2920
Formula Weight 149.03
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-3-methyl-2-butene (stabilized with Silver chip)
EINECSNumber (2)-0067&(2)-2368
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.