Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-3-phenylpropane, 98%

Catalog No. p-76308
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107290050 5 mL Glass bottle
AC107291000 100 mL Glass bottle
AC107295000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC107290050 Supplier Thermo Scientific Chemicals Supplier No. 107290050
Only null left
Add to Cart

CAS: 637-59-2 | C9H11Br | 199.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-59-2
Molecular Formula C9H11Br
Molecular Weight (g/mol) 199.09
MDL Number MFCD00000257
InChI Key XMZQWZJMTBCUFT-UHFFFAOYSA-N
Synonym 3-bromopropyl benzene, 1-bromo-3-phenylpropane, 3-phenylpropyl bromide, benzene, 3-bromopropyl, 3-bromo-1-phenylpropane, 3-bromoprop-1-yl benzene, unii-6vx623qn9v, .gamma.-bromopropyl benzene, 4-bromopropylbenzene, benzene, bromopropyl
PubChem CID 12503
IUPAC Name 3-bromopropylbenzene
SMILES BrCCCC1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.3100g/mL
Boiling Point 219.0°C to 220.0°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5450 to 1.5470
Linear Formula C6H5(CH2)3Br
Quantity 5 mL
Beilstein 05, 391
Specific Gravity 1.31
Solubility Information Solubility in water: insoluble
Formula Weight 199.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Bromo-3-phenylpropane, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-294-7
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.