Learn More
Sigma Aldrich 1-Bromo-4-(benzyloxy)-2,3-difluorobenzene
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Chloroacetone undergoes direct aldol reaction with 4-nitrobenzaldehyde catalyzed by L-proline amides
- It reacts with carboxylic acids to form ester derivatives.chloroacetone was used in microbial asymmetric reduction of chloracetone to synthesize chiral 1,2-epoxy propane
- It was used in analysis of acids by gas chromatography
- It was also used in the syntheis of methylthiazole derivatives by reacting with 1-((3-(naphtho[2,1-b]furan-2-yl)-1-phenyl-1h-pyrazol-4-yl) methylene)-2-phenylhydrazone.prices for bulk quantities on request
Specifications
Specifications
| CAS | 941294-52-6 |
| Molecular Formula | C13H9BrF2O |
| Linear Formula | C13H9BrF2O |
| MDL Number | MFCD09475823 |
| Quantity | 1 kg |
| Molecular Weight (g/mol) | 299.11 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.