Learn More
Medchemexpress LLC 1-Bromo-4-(bromomethyl)benzene-d6 | 229948-68-9 | 99.6% | C7D6Br2 | 50 MG
SDP
Catalog No. 5000433040
Encompass_Preferred
1-Bromo-4-(bromomethyl)benzene-d6 is the deuterium labeled 1-Bromo-4-(bromomethyl)benzene. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be utilized as a tracer in various studies.
  • Functions as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS.
  • Incorporation of stable heavy isotopes can aid in quantitation during drug development processes.
  • Deuteration offers the potential to influence the pharmacokinetic and metabolic profiles of pharmaceutical compounds.
Catalog No. 50-004-33040 Supplier Medchemexpress LLC Supplier No. HY79779S50MG
1-Bromo-4-(bromomethyl)benzene-d6 is the deuterium labeled 1-Bromo-4-(bromomethyl)benzene. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be utilized as a tracer in various studies.
  • Functions as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS.
  • Incorporation of stable heavy isotopes can aid in quantitation during drug development processes.
  • Deuteration offers the potential to influence the pharmacokinetic and metabolic profiles of pharmaceutical compounds.