Learn More
Medchemexpress LLC 1-bromo-4-(bromomethyl)benzene-d6 | 229948-68-9 | MFCD22565819 | 99.6% | 255.97 | C7D6Br2 | 10 MG
SDP
Catalog No. 5000299108
Encompass_Preferred
1-Bromo-4-(bromomethyl)benzene-d6 is a deuterium-labeled benzyl bromide supplied as a powder for research use. It is used as an isotopically labeled standard and synthetic intermediate in organic and medicinal chemistry, offering high purity for analytical and synthetic workflows.
  • Deuterium-labeled benzyl bromide for use as an isotopically labeled standard.
  • Molecular formula C7D6Br2 and molecular weight 255.97.
  • High purity (99.56%) suitable for analytical applications.
  • Supplied as a powder with recommended cold storage for stability.
  • Available in small milligram quantities for research-scale use.
Catalog No. 50-002-99108 Supplier Medchemexpress LLC Supplier No. HY79779S10MG
1-Bromo-4-(bromomethyl)benzene-d6 is a deuterium-labeled benzyl bromide supplied as a powder for research use. It is used as an isotopically labeled standard and synthetic intermediate in organic and medicinal chemistry, offering high purity for analytical and synthetic workflows.
  • Deuterium-labeled benzyl bromide for use as an isotopically labeled standard.
  • Molecular formula C7D6Br2 and molecular weight 255.97.
  • High purity (99.56%) suitable for analytical applications.
  • Supplied as a powder with recommended cold storage for stability.
  • Available in small milligram quantities for research-scale use.