Learn More
Medchemexpress LLC 1-Bromo-4-(bromomethyl)benzene-d6 (α,4-Dibromotoluene-d6) | 229948-68-9 | 99.6% | C7D6Br2 | 25 MG
SDP
Catalog No. 5000358270
Encompass_Preferred
1-Bromo-4-(bromomethyl)benzene-d6 is a deuterium labeled compound, useful as a tracer or internal standard for quantitative analysis. Deuteration can influence pharmacokinetic and metabolic profiles of drugs, making this product valuable for drug development research.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs
  • For research use only
Catalog No. 50-003-58270 Supplier Medchemexpress LLC Supplier No. 25MGHY79779S
1-Bromo-4-(bromomethyl)benzene-d6 is a deuterium labeled compound, useful as a tracer or internal standard for quantitative analysis. Deuteration can influence pharmacokinetic and metabolic profiles of drugs, making this product valuable for drug development research.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs
  • For research use only