Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-4-chloro-2-fluorobenzene, 98%

Catalog No. AAA1957014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1957014 25 g
AAA1957006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1957014 Supplier Thermo Scientific Chemicals Supplier No. A1957014
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1996-29-8 | C6H3BrClF | 209.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1996-29-8
Molecular Formula C6H3BrClF
Molecular Weight (g/mol) 209.44
MDL Number MFCD00079708
InChI Key FPNVMCMDWZNTEU-UHFFFAOYSA-N
Synonym 4-chloro-2-fluorobromobenzene, 2-fluoro-4-chlorobromobenzene, 2-bromo-5-chlorofluorobenzene, 1-bromo-4-chloro-2-fluoro-benzene, 1-bromo-2-fluoro-4-chlorobenzene, 2-bromo-5-chloro-1-fluorobenzene, benzene, 1-bromo-4-chloro-2-fluoro, pubchem3209, acmc-1bovk, 2-bromo-5-chlorofluorbenzene
PubChem CID 137275
IUPAC Name 1-bromo-4-chloro-2-fluorobenzene
SMILES FC1=CC(Cl)=CC=C1Br

Specifications

Density 1.678
Boiling Point 70°C to 72°C (11 mmHg)
Flash Point 92°C (197°F)
Refractive Index 1.554
Quantity 25 g
Beilstein 2081083
Formula Weight 209.45
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-chloro-2-fluorobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.