Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-4-isopropylbenzene, 97%

Catalog No. AAA1337009
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1337009 10 g
AAA1337006 5 g
AAA1337018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1337009 Supplier Thermo Scientific Chemicals Supplier No. A1337009
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 586-61-8 | C9H11Br | 199.09 g/mol

It employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It employed as a intermediate for pharmaceutical.

Solubility
Insoluble in water

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 586-61-8
Molecular Formula C9H11Br
Molecular Weight (g/mol) 199.09
MDL Number MFCD00039159
InChI Key MOZHUOIQYVYEPN-UHFFFAOYSA-N
Synonym 1-bromo-4-isopropylbenzene, 4-bromocumene, 4-bromoisopropylbenzene, p-bromocumene, 4-isopropylbromobenzene, benzene, 1-bromo-4-1-methylethyl, p-bromoisopropylbenzene, 2-p-bromophenyl propane, cumene, p-bromo, 2-4-bromophenyl propane
PubChem CID 11462
IUPAC Name 1-bromo-4-propan-2-ylbenzene
SMILES CC(C)C1=CC=C(Br)C=C1

Specifications

Melting Point -12°C
Density 1.315
Boiling Point 219°C
Flash Point >100°C (212°F)
Refractive Index 1.536
Quantity 10 g
UN Number UN3082
Beilstein 1858096
Solubility Information Insoluble in water
Formula Weight 199.1
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-isopropylbenzene
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 209-577-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.